2-(1-Methoxy)propyl acetate, 99%, Thermo Scientific Chemicals
2-(1-Methoxy)propyl acetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(1-Methoxy)propyl acetate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Packaging:
View all
Plastic drum
Catalog number 413900050
Price (USD)/ Each
216.65
Online exclusive
240.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
216.65
Online exclusive
240.00 
Save 23.35 (10%)
Add to cart
2-(1-Methoxy)propyl acetate, 99%, Thermo Scientific Chemicals
Catalog number413900050
Price (USD)/ Each
216.65
Online exclusive
240.00 
Save 23.35 (10%)
-
Add to cart
Chemical Identifiers
CAS108-65-6
IUPAC Name1-methoxypropan-2-yl acetate
Molecular FormulaC6H12O3
InChI KeyLLHKCFNBLRBOGN-UHFFFAOYNA-N
SMILESCOCC(C)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Refractive index1.4010 to 1.4030 (20°C, 589 nm)
Appearance (Form)Liquid
Water=<0.05 %
Acidity (CH3COOH)=<0.02 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only