Thermo Scientific Chemicals

Methyl 3-aminobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 413950250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 3-aminobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 413950250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 413950250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4518-10-9
IUPAC Name
methyl 3-aminobenzoate
Molecular Formula
C8H9NO2
InChI Key
VZDNXXPBYLGWOS-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC(N)=C1
Appearance (Color)
Light beige to brown
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals
Melting point
49°C to 54°C
Titration with HClO4
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text