Barnstead™ MegaPure™ Bottles
Barnstead™ MegaPure™ Bottles
Actual product may vary
Thermo Scientific™
Barnstead™ MegaPure™ Bottles
Safely store distilled water using plastic or glass Thermo Scientific™ Barnstead™ MegaPure™ Bottles compatible with Barnstead Mega-Pure Glass Stills.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
4139646L (1.62 gal.), Polyethylene Bottle
410164TS
also known as 410164
45L (11.2 gal.), Glass Bottle
41393413L (3.5 gal.), Glass Botttle
3 Options
Catalog number 413964
Price (USD)/ Each
-
Request A Quote
Description:
6L (1.62 gal.), Polyethylene Bottle
Safely store distilled water using plastic or glass Thermo Scientific™ Barnstead™ MegaPure™ Bottles compatible with Barnstead Mega-Pure Glass Stills. Water bottles for this distillation system are available in varying capacities.
Specifications
MaterialPolyethylene
For Use With (Equipment)MegaPure Still MP-1, MP-3
Description6L (1.62 gal.), Polyethylene Bottle
Capacity (English)1.62 gal.
Capacity (Metric)6 L
Unit SizeEach