Thermo Scientific Chemicals

Methyl benzenesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: 413990250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl benzenesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: 413990250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 413990250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
80-18-2
IUPAC Name
methyl benzenesulfonate
Molecular Formula
C7H8O3S
InChI Key
CZXGXYBOQYQXQD-UHFFFAOYSA-N
SMILES
COS(=O)(=O)C1=CC=CC=C1
Appearance (Color)
Clear colorless to light brown
Refractive index
1.5150 to 1.5180 (20°C, 589 nm)
Appearance (Form)
Liquid
HPLC
>=98.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text