Informational:Discover the future of Gibco Media selection with our upcoming updated experience. View details >
Gibco™

Insulin-Transferrin-Selenium (ITS -G) (100X)

Catalog number: 41400045
Have Questions?
Gibco™

Insulin-Transferrin-Selenium (ITS -G) (100X)

Catalog number: 41400045
Have Questions?
Catalog Number
41400045
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4140004510 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Gibco™ Insulin-Transferrin-Selenium (ITS-G) is used as a basal medium supplement in order to reduce the amount of fetal bovine serum (FBS) needed to culture cells. Originally used with RPMI 1640 and Minimum Essential Medium (MEM), ITS-G solution is now routinely used with other basal media to support a wide variety of cell types in the presence of 2–4% FBS.

Insulin promotes glucose and amino acid uptake, lipogenesis, intracellular transport, and the synthesis of proteins and nucleic acids. Transferrin is an iron carrier and it may also help to reduce toxic levels of oxygen radicals and peroxide. Selenium, as sodium selenite, is a co-factor for glutathione peroxidase and other proteins, and is used as an anti-oxidant in media.

The complete formulation is available.

cGMP Manufacturing and Quality System
Gibco™ ITS-G is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Mammalian Cells
Concentration
100 X
Form
Liquid
Product Type
Transferrin
Shelf Life
18 Months
Sterility
Sterile-filtered
Without Additives
No Phenol Red
Quantity
10 mL
Shipping Condition
Room Temperature

Contents & Storage

Storage conditions: 2°C to 8°C
Shipping conditions: Room temperature
Shelf life: 18 months from date of manufacture

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text