3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride Monohydrate, 98%, Thermo Scientific Chemicals
3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride Monohydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride Monohydrate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 414022500
Price (USD)/ Each
1,524.65
Online exclusive
1694.00 
Save 169.35 (10%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
1,524.65
Online exclusive
1694.00 
Save 169.35 (10%)
Add to cart
3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride Monohydrate, 98%, Thermo Scientific Chemicals
Catalog number414022500
Price (USD)/ Each
1,524.65
Online exclusive
1694.00 
Save 169.35 (10%)
-
Add to cart
Chemical Identifiers
CAS38894-11-0
IUPAC Name(2Z)-2-hydrazinylidene-3-methyl-2,3-dihydro-1,3-benzothiazole
Molecular FormulaC8H9N3S
InChI KeyPHOLIFLKGONSGY-NTMALXAHSA-N
SMILESCN1C(SC2=CC=CC=C12)=NN
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental Analysis40.1 to 42.1 % (C)
Appearance (Form)Crystalline powder
HPLC>=97.5 %
Appearance (Color)White to beige
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only