Methyl butyrate, 99%
Methyl butyrate, 99%
Methyl butyrate, 99%
Methyl butyrate, 99%
Thermo Scientific Chemicals

Methyl butyrate, 99%

CAS: 623-42-7 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
250 g
Packaging:
Glass bottle
Catalog number 414141000
Price (USD)
33.65
Special offer
Online exclusive
Ends: 26-Sep-2025
41.90
Save 8.25 (20%)
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 26-Sep-2025
41.90
Save 8.25 (20%)
Each
Add to cart
Chemical Identifiers
CAS623-42-7
IUPAC Namemethyl butanoate
Molecular FormulaC5H10O2
InChI KeyUUIQMZJEGPQKFD-UHFFFAOYSA-N
SMILESCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.3850 to 1.3900 (20°C, 589 nm)
Density(20°C) 0.893 to 0.899 g/ml
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection