Thermo Scientific Chemicals

Methyl butyrate, 99%, Thermo Scientific Chemicals

Catalog number: 414142500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl butyrate, 99%, Thermo Scientific Chemicals

Catalog number: 414142500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 414142500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
623-42-7
IUPAC Name
methyl butanoate
Molecular Formula
C5H10O2
InChI Key
UUIQMZJEGPQKFD-UHFFFAOYSA-N
SMILES
CCCC(=O)OC
Appearance (Form)
Liquid
GC
>=98.5 %
Refractive index
1.3850 to 1.3900 (20°C, 589 nm)
Density
(20°C) 0.893 to 0.899 g/ml
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text