DL-2-Methylbutyryl chloride, 98%, Thermo Scientific Chemicals
DL-2-Methylbutyryl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-2-Methylbutyryl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number 414151000
Price (USD)/ Each
498.00
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
498.00
Add to cart
DL-2-Methylbutyryl chloride, 98%, Thermo Scientific Chemicals
Catalog number414151000
Price (USD)/ Each
498.00
-
Add to cart
Chemical Identifiers
IUPAC Name2-methylbutanoyl chloride
Molecular FormulaC5H9ClO
InChI KeyXRPVXVRWIDOORM-UHFFFAOYNA-N
SMILESCCC(C)C(Cl)=O
Molecular Weight (g/mol)120.58
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Titration RCOCl>=97.5 %
Refractive index1.4160 to 1.4180 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only