Thermo Scientific Chemicals

Methyl alpha-chloroacrylate, 98+%, stabilized with hydroquinone, Thermo Scientific Chemicals

Catalog number: 414160250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl alpha-chloroacrylate, 98+%, stabilized with hydroquinone, Thermo Scientific Chemicals

Catalog number: 414160250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 414160250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
80-63-7
IUPAC Name
methyl 2-chloroprop-2-enoate
Molecular Formula
C4H5ClO2
InChI Key
AWJZTPWDQYFQPQ-UHFFFAOYSA-N
SMILES
COC(=O)C(Cl)=C
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
GC
>=98.0 %
Refractive index
1.4420 to 1.4440 (20°C, 589 nm)
Stabilizer
About 0.06% Hydroquinone

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text