Thermo Scientific Chemicals

Methyl dimethoxyacetate, 98%, Thermo Scientific Chemicals

Catalog number: 414180250
25 g, Each
Thermo Scientific Chemicals

Methyl dimethoxyacetate, 98%, Thermo Scientific Chemicals

Catalog number: 414180250
25 g, Each
Quantity
Catalog number: 414180250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
89-91-8
IUPAC Name
methyl 2,2-dimethoxyacetate
Molecular Formula
C5H10O4
InChI Key
NZTCVGHPDWAALP-UHFFFAOYSA-N
SMILES
COC(OC)C(=O)OC
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4040 to 1.4070 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text