Thermo Scientific Chemicals

Methylene Blue, pure, certified

Catalog number: 414241000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Methylene Blue, pure, certified

Catalog number: 414241000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 414241000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7220-79-3
IUPAC Name
3,7-bis(dimethylamino)-5λ⁴-phenothiazin-5-ylium trihydrate chloride
Molecular Formula
C16H24ClN3O3S
InChI Key
XQAXGZLFSSPBMK-UHFFFAOYSA-M
SMILES
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
Appearance (Color)
Dark green
Appearance (Form)
Crystalline powder
UV Lambda max
663 to 667 nm (in water)
BSC Certification
Passes
Absorption Ratio P-15/P+15
1.21 to 1.70

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text