Chemical Identifiers
IUPAC Name7-(dimethylamino)-N,N-dimethyl-3H-phenothiazin-3-iminium chloride
Molecular FormulaC16H18ClN3S
InChI KeyCXKWCBBOMKCUKX-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C
Molecular Weight (g/mol)319.85
View more