Thermo Scientific Chemicals

Methyl formate, 98%, for spectroscopy, Thermo Scientific Chemicals

Catalog number: 414340025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Methyl formate, 98%, for spectroscopy, Thermo Scientific Chemicals

Catalog number: 414340025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 414340025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
107-31-3
IUPAC Name
methyl formate
Molecular Formula
C2H4O2
InChI Key
TZIHFWKZFHZASV-UHFFFAOYSA-N
SMILES
COC=O
Color scale
=<10 APHA
UV
at 260 nm A: =<1
UV
at 320 nm A: =<0.009
Infrared spectrum
Conforms
UV
(neat)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

for UV/VIS spectroscopy
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text