Methyl formate, 98%, for spectroscopy
Methyl formate, 98%, for spectroscopy
Methyl formate, 98%, for spectroscopy
Methyl formate, 98%, for spectroscopy
Methyl formate, 98%, for spectroscopy
Thermo Scientific Chemicals

Methyl formate, 98%, for spectroscopy

CAS: 107-31-3 | C2H4O2 | 60.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 414345000
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
In stock
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS107-31-3
IUPAC Namemethyl formate
Molecular FormulaC2H4O2
InChI KeyTZIHFWKZFHZASV-UHFFFAOYSA-N
SMILESCOC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
UVat 260 nm A: =<1
UVat 320 nm A: =<0.009
Infrared spectrumConforms
UV(neat)
View more
Ask our AI about this Product

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

for UV/VIS spectroscopy
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471250Certificate of AnalysisMay 17, 2025414345000, 414340025, 414340050
A0468300Certificate of AnalysisJan 31, 2025414345000, 414340025, 414340050
A0433410Certificate of AnalysisSep 09, 2024414345000, 414340025, 414340050
A0462832Certificate of AnalysisMay 17, 2024414345000, 414340025, 414340050
A0461125Certificate of AnalysisMar 15, 2024414345000, 414340025, 414340050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection