Thermo Scientific Chemicals

Methyl glycolate, 98%, Thermo Scientific Chemicals

Catalog number: 414370250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl glycolate, 98%, Thermo Scientific Chemicals

Catalog number: 414370250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 414370250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
96-35-5
IUPAC Name
methyl 2-hydroxyacetate
Molecular Formula
C3H6O3
InChI Key
GSJFXBNYJCXDGI-UHFFFAOYSA-N
SMILES
COC(=O)CO
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Refractive index
1.4150 to 1.4180 (20°C, 589 nm)
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text