Methyl Green, zinc chloride salt, pure, certified
Methyl Green, zinc chloride salt, pure, certified
Thermo Scientific Chemicals

Methyl Green, zinc chloride salt, pure, certified

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 414381000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7114-03-6
IUPAC Namezinc(2+) bis[4-(dimethylamino)phenyl][4-(ethyldimethylazaniumyl)phenyl]methylium tetrachloride
Molecular FormulaC27H35Cl4N3Zn
InChI KeyZKOGXCSOLMZRAZ-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Zn++].CC[N+](C)(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
BSC CertificationPasses
Appearance (Color)Dark green to dark brown-red
Appearance (Form)Powder
Infrared spectrumConforms
Dye content>=80 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only