Thermo Scientific Chemicals

Methyl malonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 414510250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl malonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 414510250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 414510250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
37517-81-0
IUPAC Name
methyl 3-chloro-3-oxopropanoate
Molecular Formula
C4H5ClO3
InChI Key
UTBCRHAMJFMIIR-UHFFFAOYSA-N
SMILES
COC(=O)CC(Cl)=O
Infrared spectrum
Conforms
Refractive index
1.4310 to 1.4350 (20°C, 589 nm)
Appearance (Color)
Clear yellow
Appearance (Form)
Liquid
Titration RCOCl
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text