Methyl octanoate, 99%, Thermo Scientific Chemicals
Methyl octanoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl octanoate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 414620250
Price (USD)/ Each
47.65
Online exclusive
52.30 
Save 4.65 (9%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
47.65
Online exclusive
52.30 
Save 4.65 (9%)
Add to cart
Methyl octanoate, 99%, Thermo Scientific Chemicals
Catalog number414620250
Price (USD)/ Each
47.65
Online exclusive
52.30 
Save 4.65 (9%)
-
Add to cart
Chemical Identifiers
CAS111-11-5
IUPAC Namemethyl octanoate
Molecular FormulaC9H18O2
InChI KeyJGHZJRVDZXSNKQ-UHFFFAOYSA-N
SMILESCCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=98.5 %
Infrared spectrumConforms
Refractive index1.4165 to 1.4185 (20°C, 589 nm)
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only