Thermo Scientific Chemicals

Methyl oleate, technical, Thermo Scientific Chemicals

Catalog number: 414645000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl oleate, technical, Thermo Scientific Chemicals

Catalog number: 414645000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 414645000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67762-38-3
IUPAC Name
methyl octadec-9-enoate
Molecular Formula
C19H36O2
InChI Key
QYDYPVFESGNLHU-ZHACJKMWSA-N
SMILES
CCCCCCCCC=CCCCCCCCC(=O)OC
Appearance (Color)
Clear yellow to amber
Appearance (Form)
Viscous liquid
Infrared spectrum
Conforms
GC
>=55 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text