Thermo Scientific Chemicals

Methyl orange

Catalog number: 414661000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl orange

Catalog number: 414661000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 414661000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
547-58-0
IUPAC Name
4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
Molecular Formula
C14H14N3O3S
InChI Key
IETWCRRCPURZOC-UHFFFAOYSA-M
SMILES
CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O
Appearance (Color)
Orange
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Clarity of solution
Passes test
Visual transition interval
From pH 3.2 (pink or red) to pH 4.4 (yellow)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text