Chemical Identifiers
CAS62758-15-0
IUPAC Namedisodium 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonate 4-{[4-(ethylamino)-3-methylphenyl][(1E)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate
Molecular FormulaC39H41N5Na2O9S3
InChI KeyQZATZDKHJDOZFD-DGVIJJPGSA-L
SMILES[Na+].[Na+].CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O.CCNC1=CC=C(C=C1C)C(=C1\C=CC(=NCC)C(C)=C1)\C1=CC=C(C=C1S(O)(=O)=O)S([O-])(=O)=O
View more