Thermo Scientific Chemicals

4-Methylphthalic anhydride, 96%, Thermo Scientific Chemicals

Catalog number: 414791000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Methylphthalic anhydride, 96%, Thermo Scientific Chemicals

Catalog number: 414791000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 414791000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19438-61-0
IUPAC Name
5-methyl-1,3-dihydro-2-benzofuran-1,3-dione
Molecular Formula
C9H6O3
InChI Key
ZOXBWJMCXHTKNU-UHFFFAOYSA-N
SMILES
CC1=CC=C2C(=O)OC(=O)C2=C1
Appearance (Color)
White to yellow to light orange
Appearance (Form)
Crystalline powder or flakes
Infrared spectrum
Conforms
Melting point
88°C to 93°C
Titration by coupling
>=95 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text