Thermo Scientific Chemicals

Methylsulfuric acid sodium salt, 99%, pract., Thermo Scientific Chemicals

Catalog number: 414900250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Methylsulfuric acid sodium salt, 99%, pract., Thermo Scientific Chemicals

Catalog number: 414900250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 414900250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
512-42-5
IUPAC Name
sodium methyl sulfate
Molecular Formula
CH3NaO4S
InChI Key
DZXBHDRHRFLQCJ-UHFFFAOYSA-M
SMILES
[Na+].COS([O-])(=O)=O
Titration with NaOH
>=98.5 %
Appearance (Color)
White
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text