Thermo Scientific Chemicals

Methyl p-toluate, 99%, Thermo Scientific Chemicals

Catalog number: 414960025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Methyl p-toluate, 99%, Thermo Scientific Chemicals

Catalog number: 414960025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 414960025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
99-75-2
IUPAC Name
methyl 4-methylbenzoate
Molecular Formula
C9H10O2
InChI Key
QSSJZLPUHJDYKF-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(C)C=C1
Appearance (Color)
Clear colorless to light yellow
Melting point
32°C to 35°C
GC
>=98.5 %
Appearance (Form)
Liquid or solid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text