Methyl p-toluate, 99%, Thermo Scientific Chemicals
Methyl p-toluate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl p-toluate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
100 g
2.5 kg
Packaging:
View all
Plastic drum
Catalog number 414960100
Price (USD)/ Each
-
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS99-75-2
IUPAC Namemethyl 4-methylbenzoate
Molecular FormulaC9H10O2
InChI KeyQSSJZLPUHJDYKF-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Melting point32°C to 35°C
GC>=98.5 %
Appearance (Form)Liquid or solid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only