Thermo Scientific Chemicals

Methyl Violet 2B, Thermo Scientific Chemicals

Catalog number: 415070250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl Violet 2B, Thermo Scientific Chemicals

Catalog number: 415070250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 415070250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
8004-87-3
IUPAC Name
bis[4-(dimethylamino)phenyl][4-(methylamino)phenyl]methylium
Molecular Formula
C24H28N3
InChI Key
HJPVVBCICYOONZ-UHFFFAOYSA-N
SMILES
CNC1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
Dye content
>=75 %
UV Lambda max
582 to 588 nm
Absorption Ratio P-15/P+15
1.06 to 1.19
BSC Certification
Passes
Appearance (Color)
Green

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text