Thermo Scientific Chemicals

Mineral oil, pure, Thermo Scientific Chemicals

Catalog number: 415080010
1 L, Each
Thermo Scientific Chemicals

Mineral oil, pure, Thermo Scientific Chemicals

Catalog number: 415080010
1 L, Each
Quantity
Catalog number: 415080010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
8042-47-5
Infrared spectrum
Conforms
Refractive index
1.4730 to 1.4800 (20°C, 589 nm)
Density
(15°C) 0.860 to 0.880 g/ml
Viscosity
63 to 80 mm²/s (40°C)
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text