Chemical Identifiers
CAS18378-89-7
IUPAC Name(2S,3S)-3-[(1S,3S,4R)-3,4-dihydroxy-1-methoxy-2-oxopentyl]-2-{[(2S,4R,5R,6R)-4-{[(2S,4R,5R,6R)-4-{[(2S,4S,5R,6R)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy}-5-hydroxy-6-methyloxan-2-yl]oxy}-5-hydroxy-6-methyloxan-2-yl]oxy}-6-{[(2S,4R,5R,6R)-4-{[(2S,4R,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy}-5-hydroxy-6-methyloxan-2-yl]oxy}-8,9-dihydroxy-7-methyl-1,2,3,4-tetrahydroanthracen-1-one
Molecular FormulaC52H76O24
InChI KeyCFCUWKMKBJTWLW-WZIVUMCRSA-N
SMILESCO[C@@H]([C@@H]1CC2=C(C(=O)[C@H]1O[C@H]1C[C@@H](O[C@H]3C[C@@H](O[C@H]4C[C@](C)(O)[C@H](O)[C@@H](C)O4)[C@H](O)[C@@H](C)O3)[C@H](O)[C@@H](C)O1)C(O)=C1C(O)=C(C)C(O[C@H]3C[C@@H](O[C@H]4C[C@@H](O)[C@H](O)[C@@H](C)O4)[C@H](O)[C@@H](C)O3)=CC1=C2)C(=O)[C@@H](O)[C@@H](C)O
View more