Thermo Scientific Chemicals

Naphthenic acids, pract., Thermo Scientific Chemicals

Catalog number: 415281000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Naphthenic acids, pract., Thermo Scientific Chemicals

Catalog number: 415281000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 415281000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1338-24-5
IUPAC Name
3-(3-ethylcyclopentyl)propanoic acid
Molecular Formula
C10H18O2
InChI Key
WVRFSLWCFASCIS-UHFFFAOYNA-N
SMILES
CCC1CCC(CCC(O)=O)C1
Appearance (Color)
Clear colorless to yellow to brown
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Density
(20°C) 910 to 940 g/l
Acid value
230 to 260 mg KOH/g

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text