Chemical Identifiers
CAS1919-91-1
IUPAC Namedisodium 6-bromo-3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-yl phosphate
Molecular FormulaC18H13BrNNa2O6P
InChI KeyHESHHBAVKCAKQF-UHFFFAOYSA-L
SMILES[Na+].[Na+].COC1=CC=CC=C1NC(=O)C1=C(OP([O-])([O-])=O)C=C2C=CC(Br)=CC2=C1
View more