Thermo Scientific Chemicals

1,2-Naphthoquinone-4-sulfonic acid sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 415401000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1,2-Naphthoquinone-4-sulfonic acid sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 415401000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 415401000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
521-24-4
IUPAC Name
sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulfonate
Molecular Formula
C10H5NaO5S
InChI Key
UBLXEEBHYISRFM-UHFFFAOYSA-M
SMILES
[Na+].[O-]S(=O)(=O)C1=CC(=O)C(=O)C2=CC=CC=C12
Infrared spectrum
Conforms
Titration Iodometric
>=97.5 %
Appearance (Color)
Orange
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text