Thermo Scientific Chemicals

1-Naphthyl isocyanate, 99%, Thermo Scientific Chemicals

Catalog number: 415410050
5 g, Each
Thermo Scientific Chemicals

1-Naphthyl isocyanate, 99%, Thermo Scientific Chemicals

Catalog number: 415410050
5 g, Each
Quantity
Catalog number: 415410050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86-84-0
IUPAC Name
1-isocyanatonaphthalene
Molecular Formula
C11H7NO
InChI Key
BDQNKCYCTYYMAA-UHFFFAOYSA-N
SMILES
O=C=NC1=C2C=CC=CC2=CC=C1
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.6324 to 1.6344 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text