Nickel(II) oxide, 97%, reagent, powder, Thermo Scientific Chemicals
Nickel(II) oxide, 97%, reagent, powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel(II) oxide, 97%, reagent, powder, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number 415585000
Price (USD)/ Each
210.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
248.00 
Save 37.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
210.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
248.00 
Save 37.35 (15%)
Add to cart
Nickel(II) oxide, 97%, reagent, powder, Thermo Scientific Chemicals
Catalog number415585000
Price (USD)/ Each
210.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
248.00 
Save 37.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameoxonickel
Molecular FormulaNiO
InChI KeyGNRSAWUEBMWBQH-UHFFFAOYSA-N
SMILESO=[Ni]
Molecular Weight (g/mol)74.69
View more
SpecificationsSpecification SheetSpecification Sheet
Magnesium (Mg)=<0.01 %
Manganese (Mn)=<0.001 %
Appearance (Color)Green
Appearance (Form)Powder
Titration Complexometric>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only