Thermo Scientific Chemicals

Nickel phthalocyanine, 95%, Thermo Scientific Chemicals

Catalog number: 415600250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Nickel phthalocyanine, 95%, Thermo Scientific Chemicals

Catalog number: 415600250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 415600250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14055-02-8
IUPAC Name
nickel(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
Molecular Formula
C32H16N8Ni
InChI Key
LVIYYTJTOKJJOC-UHFFFAOYSA-N
SMILES
[Ni++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
Appearance (Color)
Dark blue
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Gravimetry
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text