Nickel(II) sulfate hexahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Nickel(II) sulfate hexahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel(II) sulfate hexahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number 415610010
Price (USD)/ Each
252.65
Online exclusive
281.00 
Save 28.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
252.65
Online exclusive
281.00 
Save 28.35 (10%)
Add to cart
Nickel(II) sulfate hexahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Catalog number415610010
Price (USD)/ Each
252.65
Online exclusive
281.00 
Save 28.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10101-97-0
IUPAC Namenickel(2+) hexahydrate sulfate
Molecular FormulaH12NiO10S
InChI KeyRRIWRJBSCGCBID-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue-green
Appearance (Form)Crystals
Titration Complexometric98.0 to 102.0 %
Insoluble matter=<0.005 %
Nitrogen compounds (as N)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only