Nicotine ditartrate dihydrate, 98%
Nicotine ditartrate dihydrate, 98%
Nicotine ditartrate dihydrate, 98%
Nicotine ditartrate dihydrate, 98%
Nicotine ditartrate dihydrate, 98%
Thermo Scientific Chemicals

Nicotine ditartrate dihydrate, 98%

CAS: 6019-06-3 | C18H30N2O14 | 498.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 415660100
Price (USD)
80.65
Online Exclusive
89.90
Save 9.25 (10%)
Each
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
80.65
Online Exclusive
89.90
Save 9.25 (10%)
Each
Add to cart
Chemical Identifiers
CAS6019-06-3
IUPAC Namebis((2R,3R)-2,3-dihydroxybutanedioic acid) 3-[(2S)-1-methylpyrrolidin-2-yl]pyridine dihydrate
Molecular FormulaC18H30N2O14
InChI KeyLDMPZNTVIGIREC-HYRPJEHINA-N
SMILESO.O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCC[C@H]1C1=CN=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Water5 to 10 % (K.F.)
Titration with TBAH>=97.5 % (On dry substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link