Chemical Identifiers
IUPAC Name[9-(diethylamino)benzo[a]phenoxazin-5-ylidene]azanium;sulfate
Molecular FormulaC40H40N6O6S
InChI KeyQIRDPEPUXNCOLD-UHFFFAOYSA-N
SMILESCCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.[O-]S(=O)(=O)[O-]
Molecular Weight (g/mol)732.84
View more