Nile Blue A
Thermo Scientific Chemicals

Nile Blue A

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 415690500
Price (USD)/ Each
143.65
Online exclusive
159.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
143.65
Online exclusive
159.00 
Save 15.35 (10%)
Add to cart
Nile Blue A
Catalog number415690500
Price (USD)/ Each
143.65
Online exclusive
159.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Name[9-(diethylamino)benzo[a]phenoxazin-5-ylidene]azanium;sulfate
Molecular FormulaC40H40N6O6S
InChI KeyQIRDPEPUXNCOLD-UHFFFAOYSA-N
SMILESCCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.[O-]S(=O)(=O)[O-]
Molecular Weight (g/mol)732.84
View more
SpecificationsSpecification SheetSpecification Sheet
UV Lambda max635 to 646 nm
Absorption Ratio P-15/P+151.01 to 1.16
BSC CertificationPasses
Appearance (Color)Dark green to black
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only