Nile Red, 99%, pure
Nile Red, 99%, pure
Nile Red, 99%, pure
Thermo Scientific Chemicals

Nile Red, 99%, pure

CAS: 7385-67-3 | C20H18N2O2 | 318.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Packaging:
Glass bottle
Catalog number 415711000
Price (USD)
106.65
Online Exclusive
118.00
Save 11.35 (10%)
Each
-
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
106.65
Online Exclusive
118.00
Save 11.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7385-67-3
IUPAC Name8-(diethylamino)-12H-10-oxa-5-azatetraphen-12-one
Molecular FormulaC20H18N2O2
InChI KeyVOFUROIFQGPCGE-UHFFFAOYSA-N
SMILESCCN(CC)C1=CC=C2N=C3C(OC2=C1)=CC(=O)C1=CC=CC=C31
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to dark red to brown
Infrared spectrumConforms
UV spectrumComplies
UV spectrophotometry>=98.5 %
Appearance (Form)Powder
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473601Certificate of AnalysisJun 27, 2025415710010, 415711000
A0469714Certificate of AnalysisMay 17, 2025415710010, 415711000
A0457724Certificate of AnalysisNov 23, 2023415710010, 415711000
A0453709Certificate of AnalysisAug 25, 2023415710010, 415711000
A0450812Certificate of AnalysisMay 02, 2023415710010, 415711000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link