Thermo Scientific Chemicals

Ninhydrin, Reagent ACS

Catalog number: 415720250
25 g, Glass Bottle, Each
Thermo Scientific Chemicals

Ninhydrin, Reagent ACS

Catalog number: 415720250
25 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 415720250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
485-47-2
IUPAC Name
2,2-dihydroxy-2,3-dihydro-1H-indene-1,3-dione
Molecular Formula
C9H6O4
InChI Key
FEMOMIGRRWSMCU-UHFFFAOYSA-N
SMILES
OC1(O)C(=O)C2=CC=CC=C2C1=O
Appearance (Color)
White to light brown
Appearance (Form)
Crystals
Infrared spectrum
Conforms
Identification
Passes test
Melting point
Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text