Thermo Scientific Chemicals

Nitrilotriacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: 415740051
5 kg, Plastic drum, Each
Thermo Scientific Chemicals

Nitrilotriacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: 415740051
5 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 415740051
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
139-13-9
IUPAC Name
2-[bis(carboxymethyl)amino]acetic acid
Molecular Formula
C6H9NO6
InChI Key
MGFYIUFZLHCRTH-UHFFFAOYSA-N
SMILES
OC(=O)CN(CC(O)=O)CC(O)=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration excess NaOH
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Nitrilotriacetic acid (2,2',2-nitrilotriacetic acid) is a tricarboxylic acid used for chemical purification


Applications

  • A known mutagen, suitable for cell culture experiments of DNA damage
  • Nitrilotriacetic acid can be used for protein elution using the His-tag method and chemical chelation
  • Nitrilotriacetic acid is capable of chelating Ca2+, Co2+, Cu2+, and Fe3+
  • Intended for sensitive applications such as HPLC
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text