Thermo Scientific Chemicals

4'-Nitroacetanilide, 99%, Thermo Scientific Chemicals

Catalog number: 415750250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4'-Nitroacetanilide, 99%, Thermo Scientific Chemicals

Catalog number: 415750250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 415750250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
104-04-1
IUPAC Name
N-(4-nitrophenyl)acetamide
Molecular Formula
C8H8N2O3
InChI Key
NQRLPDFELNCFHW-UHFFFAOYSA-N
SMILES
CC(=O)NC1=CC=C(C=C1)[N+]([O-])=O
Melting point
213°C to 216°C
HPLC
>=98.5 %
Infrared spectrum
Conforms
Appearance (Form)
Solid
Appearance (Color)
Yellow to yellow-green to green-brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text