Thermo Scientific Chemicals

Nitrobenzene, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 415780025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Nitrobenzene, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 415780025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 415780025
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
98-95-3
IUPAC Name
nitrobenzene
Molecular Formula
C6H5NO2
InChI Key
LQNUZADURLCDLV-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=CC=C1
Water soluble titr. acid
=<0.0005 meq/g
Appearance (Color)
Clear yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 3
    Acute dermal toxicity
    Category 3
    Acute Inhalation Toxicity - Vapors
    Category 3
    Carcinogenicity
    Category 2
    Reproductive Toxicity
    Category 1B
    Specific target organ toxicity - (repeated exposure)
    Category 1
    Chronic aquatic toxicity
    Category 3
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H301 + H311 + H331 - Toxic if swallowed, in contact with skin or if inhaled

    H351 - Suspected of causing cancer

    H360F - May damage fertility

    H372 - Causes damage to organs through prolonged or repeated exposure

    H412 - Harmful to aquatic life with long lasting effects

    EU Specific Hazard Statements

    Combustible liquid

    Precautionary Statements

    P260 - Do not breathe dust/fume/gas/mist/vapors/spray

    P273 - Avoid release to the environment

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P310 - IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician

    P302 + P350 - IF ON SKIN: Gently wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing

    Additional EU labelling

    Restricted to professional users