Thermo Scientific Chemicals

Nitrobenzene, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 415780250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Nitrobenzene, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 415780250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 415780250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-95-3
IUPAC Name
nitrobenzene
Molecular Formula
C6H5NO2
InChI Key
LQNUZADURLCDLV-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=CC=C1
Water soluble titr. acid
=<0.0005 meq/g
Appearance (Color)
Clear yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text