Nitrobenzene, 99+%, ACS reagent, Thermo Scientific Chemicals
Nitrobenzene, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nitrobenzene, 99+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
2.5 L
Packaging:
Glass bottle
Catalog number 415785000
Price (USD)/ Each
86.65
Online exclusive
95.90 
Save 9.25 (10%)
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
86.65
Online exclusive
95.90 
Save 9.25 (10%)
Add to cart
Nitrobenzene, 99+%, ACS reagent, Thermo Scientific Chemicals
Catalog number415785000
Price (USD)/ Each
86.65
Online exclusive
95.90 
Save 9.25 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Namenitrobenzene
Molecular FormulaC6H5NO2
InChI KeyLQNUZADURLCDLV-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1
Molecular Weight (g/mol)123.11
View more
SpecificationsSpecification SheetSpecification Sheet
Water soluble titr. acid=<0.0005 meq/g
Appearance (Color)Clear yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only