Thermo Scientific Chemicals

4-Nitrophthalic acid, 99%, Thermo Scientific Chemicals

Catalog number: 416121000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Nitrophthalic acid, 99%, Thermo Scientific Chemicals

Catalog number: 416121000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 416121000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
610-27-5
IUPAC Name
4-nitrobenzene-1,2-dicarboxylic acid
Molecular Formula
C8H5NO6
InChI Key
SLBQXWXKPNIVSQ-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(C=C1C(O)=O)[N+]([O-])=O
Appearance (Color)
Yellow to beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
163°C to 167°C
Titration with TBAH
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text