4-Nitrophthalonitrile, 99%
4-Nitrophthalonitrile, 99%
4-Nitrophthalonitrile, 99%
4-Nitrophthalonitrile, 99%
Thermo Scientific Chemicals

4-Nitrophthalonitrile, 99%

CAS: 31643-49-9 | C8H3N3O2 | 173.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 416150250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS31643-49-9
IUPAC Name4-nitrobenzene-1,2-dicarbonitrile
Molecular FormulaC8H3N3O2
InChI KeyNTZMSBAAHBICLE-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(C#N)C(=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to beige to light green to light grey
Melting point140°C to 144°C
HPLC>=98.5 %
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0466167Certificate of AnalysisJan 31, 2025416150250, 416150050
A0462271Certificate of AnalysisAug 14, 2024416150250, 416150050
A0434261Certificate of AnalysisMar 22, 2024416150250, 416150050
A0371341Certificate of AnalysisDec 14, 2022416150250, 416150050
A0428519Certificate of AnalysisApr 09, 2021416150250, 416150050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection