Thermo Scientific Chemicals

1-Octanesulfonic Acid, Sodium Salt Anhydrous, HPLC Grade, Thermo Scientific Chemicals

Catalog number: 416360050
5 g, Each
Thermo Scientific Chemicals

1-Octanesulfonic Acid, Sodium Salt Anhydrous, HPLC Grade, Thermo Scientific Chemicals

Catalog number: 416360050
5 g, Each
Quantity
Catalog number: 416360050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5324-84-5
IUPAC Name
sodium octane-1-sulfonate
Molecular Formula
C8H17NaO3S
InChI Key
HRQDCDQDOPSGBR-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCCS([O-])(=O)=O
UV
at 210 nm A: =<1.00
UV
at 220 nm A: =<0.10
UV
at 254 nm A: =<0.04
UV
at 280 nm A: =<0.04
Appearance (Form)
Crystalline solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text