Thermo Scientific Chemicals

Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals

Catalog number: 416500250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals

Catalog number: 416500250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 416500250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
142-31-4
IUPAC Name
sodium octyl sulfate
Molecular Formula
C8H17NaO4S
InChI Key
WFRKJMRGXGWHBM-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCCOS([O-])(=O)=O
Appearance (Color)
White to almost white
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration after Ion exchange
98.5 to 101.5 % (on dry substance)
Loss on drying
=<2 % (105°C) (vacuum/P2O5)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ion pair chromatography
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text